EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Scope and limitations of a DMF bio-alternative within Sonogashira cross-coupling and Cacchi-type annulation.
PMID 27829904
Abstract
Pd-catalysed C-C bond formation is an essential tool within the pharmaceutical and agrochemical industries. Many of these reactions rely heavily on polar aprotic solvents; however, despite their utility, these solvents are incompatible with the drive towards more sustainable chemical synthesis. Herein, we describe the scope and limitations of an alternative to DMF derived from renewable sources (CyreneTM) in Sonogashira cross-coupling and Cacchi-type annulations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
807796
Cyrene, BioRenewable
C6H8O3