EMAIL THIS PAGE TO A FRIEND
American journal of translational research
Upregulation of long non-coding RNA LINC00152 by SP1 contributes to gallbladder cancer cell growth and tumor metastasis via PI3K/AKT pathway.
PMID 27829993
Abstract
Gallbladder cancer (GBC) is one of the most lethal cancers with poor prognosis. In this study, we report that the long non-coding RNA LINC00152 is significantly upregulated in GBC tissues and cell lines. The high LINC00152 levels correlated positively with tumor status progression, lymph node invasion and TNM stage advancement. Functionally, we revealed that LINC00152 dramatically promoted cell proliferation, metastasis and inhibited apoptosis
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU018231 MISSION® esiRNA, esiRNA targeting human SP1 (esiRNA1)