EMAIL THIS PAGE TO A FRIEND
Nature communications
Pure crystal orientation and anisotropic charge transport in large-area hybrid perovskite films.
PMID 27830694
Abstract
Controlling crystal orientations and macroscopic morphology is vital to develop the electronic properties of hybrid perovskites. Here we show that a large-area, orientationally pure crystalline (OPC) methylammonium lead iodide (MAPbI