EMAIL THIS PAGE TO A FRIEND
Scientific reports
Additive manufacturing of micrometric crystallization vessels and single crystals.
PMID 27830827
Abstract
We present an all-additive manufacturing method that is performed at mild conditions, for the formation of organic single crystals at specific locations, without any photolithography prefabrication process. The method is composed of two steps; inkjet printing of a confinement frame, composed of a water soluble electrolyte. Then, an organic semiconductor solution is printed within the confinement to form a nucleus at a specific location, followed by additional printing, which led to the growth of a single crystal. The specific geometry of the confinement enables control of the specific locations of the single crystals, while separating the nucleation and crystal growth processes. By this method, we printed single crystals of perylene, which are suitable for the formation of OFETs. Moreover, since this method is based on a simple and controllable wet deposition process, it enables formation of arrays of single crystals at specific locations, which is a prerequisite for mass production of active organic elements on flexible substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
484423
1-Phenoxy-2-propanol, ≥93%
C9H12O2