EMAIL THIS PAGE TO A FRIEND
Scientific reports
Noscapine targets EGFR
PMID 27830833
Abstract
Osteosarcoma, the most common primary malignant bone tumor, usually arises in the metaphysis of long bones. Amplification and mutation of the epidermal growth factor receptor (EGFR) gene represent signature genetic abnormalities encountered in osteosarcoma. Noscapine is a benzylisoquinoline alkaloid derived from the opium poppy Papaver somniferum. Recently several studies have suggested its anti-cancer effect in melanoma, ovarian cancer, gliomas, breast cancer, lung cancer, and colon cancer. However, the underlying molecular mechanism for its anti-cancer effect still remains unclear. In this paper, we found the mechanism of noscapine effectively suppressed proliferation and invasion of MG63 cell line by inhibiting the phosphorylation of EGFR and its downstream pathway.