EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
Cocaine sensitization increases subthreshold activity in dopamine neurons from the ventral tegmental area.
PMID 27832596
Abstract
The progressive escalation of psychomotor responses that results from repeated cocaine administration is termed sensitization. This phenomenon alters the intrinsic properties of dopamine (DA) neurons from the ventral tegmental area (VTA), leading to enhanced dopaminergic transmission in the mesocorticolimbic network. The mechanisms underlying this augmented excitation are nonetheless poorly understood. DA neurons display the hyperpolarization-activated, nonselective cation current, dubbed I This work uses a new approach that directly extracts important biophysical parameters from alpha function-evoked synaptic potentials. Two of these parameters are the cell membrane capacitance (C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8912
Cocaine free base
C17H21NO4