EMAIL THIS PAGE TO A FRIEND
Food & nutrition research
Twelve-week-conjugated linoleic acid supplementation has no effects on the selected markers of atherosclerosis in obese and overweight women.
PMID 27834186
Abstract
The antiatherogenic effect of conjugated linoleic acid (CLA) has been demonstrated in animal models. Although there are plenty of In this study, we assessed the impact of CLA supplementation on the levels of atherosclerosis markers - high-sensitivity C-reactive protein (hs-CRP) and asymmetrical dimethylarginine (ADMA). Seventy-four adult female subjects with body mass index ≥25 kg/m No significant differences were found in hs-CRP and ADMA levels before and after nutritional intervention between both groups. The changes in hs-CRP and ADMA concentration values (Δhs-CRP; ΔADMA median [interquartile range]) did not differ between subjects from the placebo (-0.1 [-0.8 to 0.3]; -0.02 [-0.12 to 0.14]) and CLA (0.2 [-0.7 to 0.9]; 0.04 [-0.14 to 0.13]) groups. The incidence of reduction of hs-CRP or ADMA concentration was not different in subjects of the CLA group compared to those of the placebo group (41.9% vs. 50%, relative risk [RR]=0.8387, 95% confidence interval [CI]=0.4887-1.4493, Twelve weeks of CLA supplementation had no effect on selected markers of atherosclerosis in obese and overweight women.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4268
NG,NG-Dimethylarginine dihydrochloride
C8H18N4O2 · 2HCl