EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Adenosine triphosphate is a critical determinant for VEGFR signal during hypoxia.
PMID 27834196
Abstract
Hypoxia induces angiogenesis through the VEGF signaling pathway; however, signal propagation of VEGF in hypoxia is not fully understood. In this study, we examined alterations in VEGF signaling during hypoxia conditions and its determinant in endothelial cells. To analyze VEGF signaling during hypoxia, human umbilical vein endothelial cells (HUVECs) were exposed to 3 h of hypoxia (1% O