EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Receptor Species-dependent Desensitization Controls KCNQ1/KCNE1 K+ Channels as Downstream Effectors of Gq Protein-coupled Receptors.
PMID 27834678
Abstract
Activation of G
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6500
Acetylcholine bromide, ≥99%
C7H16BrNO2