EMAIL THIS PAGE TO A FRIEND
Mutagenesis
Oxidant generation, DNA damage and cytotoxicity by a panel of engineered nanomaterials in three different human epithelial cell lines.
PMID 27834732
Abstract
Due to the steeply increased use of nanomaterials (NMs) for commercial and industrial applications, toxicological assessment of their potential harmful effects is urgently needed. In this study, we compared the DNA-damaging properties and concurrent cytotoxicity of a panel of 10 engineered NMs in three different cell lines in relation to their intrinsic oxidant generating properties. The human epithelial cell lines A549, HK-2 and HepG2 were chosen to represent relevant target organs for NMs in the lung, kidney and liver. Cytotoxicity, evaluated by WST-1 assay in the treatment concentration range of 0.3-80 µg/cm