EMAIL THIS PAGE TO A FRIEND
Oncotarget
Mechanisms of transcriptional activation of the stimulator of interferon genes by transcription factors CREB and c-Myc.
PMID 27835584
Abstract
Stimulator of interferon genes (STING) plays an important role in host defense, autoimmune disease, osteoclast differentiation and anti-tumor response. Although many downstream targets have been studied in depth, the regulation of STING gene expression remains largely unknown. Here we demonstrate that transcription factors CREB and c-Myc maintain the transcriptional activity of STING. By 5'-rapid amplification of cDNA ends analysis, we identified the transcriptional start site (TSS) of STING. We illustrated that the region -124/+1 relative to TSS was sufficient for full promoter activity by a series of 5' deletion promoter constructs. Transcriptional activity of the STING minimal promoter was dependent on CREB and c-Myc binding motifs and was abolished after mutation of these two DNA elements. Chromatin immunoprecipitation assays demonstrated that transcription factors CREB and c-Myc bind to STING promoter in vivo. Overexpression of CREB and c-Myc increased the STING promoter activity. Meanwhile, knocking-down of CREB and c-Myc by a small interfering RNA (siRNA) strategy markedly reduced endogenous STING expression. In summary, these results demonstrated that transcription factors CREB and c-Myc are involved in the regulation of STING transcription.