EMAIL THIS PAGE TO A FRIEND
Talanta
Reliable, rapid and simple method for the analysis of phthalates in sediments by ultrasonic solvent extraction followed by head space-solid phase microextraction gas chromatography mass spectrometry determination.
PMID 27837884
Abstract
In this work, a new reliable, simple and fast method for the determination of six PAEs in sediments, based on ultrasonic solvent extraction (USE) followed by head space solid phase microextraction (HS-SPME) and gas chromatography-mass spectrometry determination (GC-MS), is proposed. The extraction parameters were studied, and the most favourable conditions were selected. The analytical features of the method were calculated: matrix effect, accuracy (ranged from 90% to 111%), repeatability and intermediate precision (RSD <10%), detection and quantification limits of the method (ranged from 0.001µgg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274623
Dess-Martin periodinane, 97%
C13H13IO8