EMAIL THIS PAGE TO A FRIEND
Clinical oral investigations
Influence of bioactive particles on the chemical-mechanical properties of experimental enamel resin infiltrants.
PMID 27838844
Abstract
This study aimed at evaluating the chemophysical properties of experimental resin infiltrants (ERIs) doped with different bioactive particles. A control resin infiltrant (CR) was formulated using triethylene glycol dimethacrylate (TEGDMA) and ethoxylated bisphenol A dimethacrylate (BisEMA). Moreover, five experimental ERIs were also created by incorporating the following bioactive fillers (10 wt%) into the CR: hydroxyapatite (Hap), amorphous calcium phosphate (ACP), zinc-polycarboxylated bioactive glass (BAG-Zn), bioactive glass 45S5 (BAG 45S5), and calcium silicate modified with beta tricalcium phosphate (β-TCP). ICON® resin infiltrant was also used as control. All the ERIs used in this study were assessed for degree of conversion (DC), Knoop microhardness (KHN), softening ratio (SR), tensile cohesive strength (TCS), modulus of elasticity (E-modulus), water sorption (WS), and solubility (SL). Data were subjected to ANOVA and Tukey's test (α = 5%). ICON® presented the lowest DC, KHN, TCS, E-modulus, and SR. Incorporation of bioactive fillers into CR caused significant increase in the KHN. Conversely, no significant effect was observed on DC, TCS, and E-modulus. The resin infiltrant containing Hap showed a significant increase in softening ratio, while, ICON® presented the highest WS and SL. The WS of ACP-doped resin infiltrant was significantly higher than that of the Hap-doped infiltrant. The SL of the ACP-doped infiltrant was higher than CR BAG-Zn or BAG 45S5. The incorporation of bioactive particles into experimental resin infiltrants can improve the chemomechanical properties and reduce water sorption and solubility. Resin infiltrants doped with bioactive particles may improve the long-term performance of the treatment of white-spot lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156329
Bisphenol A dimethacrylate, >98%
C23H24O4