EMAIL THIS PAGE TO A FRIEND
The journal of physiological sciences : JPS
Altered uterine contractility in response to β-adrenoceptor agonists in ovarian cancer.
PMID 27838886
Abstract
We aimed to prospectively examine β-adrenoceptor-mediated uterine contractility in women suffering from gynecological malignancies. Myometrial specimens were obtained from non-pregnant women undergoing hysterectomy for benign gynecological disorders, and ovarian, endometrial, synchronous ovarian-endometrial, and cervical cancer. Contractions of myometrial strips in an organ bath before and after cumulative dosages of β
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1385
Butoxamine hydrochloride, analytical standard, for drug analysis, mixture of diastereomers
C15H25NO3 · HCl