EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
The combination of β-caryophyllene, baicalin and catechin synergistically suppresses the proliferation and promotes the death of RAW267.4 macrophages in vitro.
PMID 27840942
Abstract
β-caryophyllene, which is a constituent of many essential oils, has been known to be a selective agonist of the cannabinoid receptor type-2 and to exert cannabimimetic anti-inflammatory effects in animals. The effects of β-caryophyllene on macrophages have not been investigated to date. This study was undertaken to determine whether β-caryophyllene exerts suppressive effects on mouse RAW267.4 macrophages inxa0vitro in comparison with other botanical molecules that exert antioxidant and anti-inflammatory effects. The proliferation of RAW267.4 cells was suppressed by culture with β-caryophyllene (50, 100 and 200xa0µg/ml of medium) or curcuminxa0(100 and 200xa0µg/ml). Culture with baicalinxa0(1-200xa0µg/ml) or (+)-catechin (1-200xa0µg/ml) did not exert an effect on RAW267.4 cells. The combination of β-caryophyllenexa0(1 or 10xa0µg/ml) with either curcuminxa0(1xa0or 10xa0µg/ml), baicalin (1 or 10xa0µg/ml) or (+)-catechin (1 or 10xa0µg/ml) did not exert any suppressive effects on cell proliferation. Of note, the combination of all three agents, β-caryophyllene, baicalin and (+)-catechin at the concentration of either 1 or 10xa0µg/ml was found to exert synergistic suppressive effects on cell proliferation. Moreover, the combination of β-caryophyllene, baicalin and (+)-catechin synergistically promoted the death of RAW267.4 cells. Such an effect was blocked by culture with caspase-3 inhibitor. The combination of the three agents decreased the protein levels of Akt, mitogen-activated protein kinasexa0(MAPK) and cyclooxygenasexa0(COX)-1 orxa0-2. On the whole, this study demonstrates that the combination of β-caryophyllene, baicalin and (+)-catechin exerts synergistic suppressive effects on macrophages inxa0vitro. This composition may be a useful as an anti-inflammatory treatment strategy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4441
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid
C6H11NOS2