EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
Curcumin based chemosensor for selective detection of fluoride and cyanide anions in aqueous media.
PMID 27841894
Abstract
The conjugate N,N-dimethyl curcumin analogue fluorophore dye 1 has been synthesized and its performance as a sensor was demonstrated. As a fluoride and cyanide sensor it enabled visual detection, and showed changes in UV-vis and fluorescence spectra in the presence of fluoride and cyanide ions in aqueous medium. The Job's plot indicated that the formation of a complex between dye-1 fluoride ions has a 1 : 1 stoichiometric ratio.