Postconditioning: "Toll-erating" mesenteric ischemia-reperfusion injury?
PMID 27842908
Postconditioning may prove to be a suitable method to decrease ischemia-reperfusion injury of intestine after mesenteric arterial occlusion. Toll-like-receptor-4 is involved in the pathophysiology of organ damage after ischemia-reperfusion; therefore, the aim of our study was to investigate the effect of postconditioning on the mucosal expression of toll-like-receptor-4. Male Wistar rats (n = 10/group) underwent 60 minutes of superior mesenteric artery occlusion followed by 6 hours of reperfusion in 3 groups: sham-operated, ischemia-reperfusion, and a postconditioned group. Postconditioning was performed by 6 alternating cycles of 10 seconds of reperfusion/reocclusion. Blood and tissue samples were collected at the end of reperfusion. Intestinal histopathologic changes and immunohistochemical expression of mucosal caspase-3, antioxidant status, and protein levels of high-mobility group box-1 and toll-like-receptor-4 were assessed. Immunofluorescent labeling and confocal microscopic analysis of toll-like-receptor-4 were performed. Mucosal and serum levels of interleukin-6 and tumor necrosis factor-α protein were measured. Histologic alterations in the postconditioned group were associated with decreased caspase-3 positivity, less toll-like-receptor-4 mRNA, and less protein expression of high-mobility group box-1 and toll-like-receptor-4 in the intestinal villi compared with the ischemia-reperfusion group. Furthermore, a significantly improved antioxidant state of the intestinal mucosa and less mucosal and serum protein levels of interleukin-6 and tumor necrosis factor-α were detected in the postconditioned group. Small intestinal ischemia-reperfusion injury in male Wistar rats caused by the occlusion of the superior mesenteric artery was ameliorated by the use of postconditioning, showing a more favorable inflammatory response, which may be attributed to the decreased mucosal expression of toll-like-receptor-4.
Related Materials