EMAIL THIS PAGE TO A FRIEND
The journal of sexual medicine
Off-Target Effect of Sildenafil on Postsurgical Erectile Dysfunction: Alternate Pathways and Localized Delivery System.
PMID 27843073
Abstract
There is no consensus on the best oral phosphodiesterase type 5 inhibitor (PDE5I) for patients undergoing penile rehabilitation after surgical nerve injury. To determine the mechanism of PDE5I on cultured neuronal cells and the effectiveness of local drug delivery using nanospheres (NSPs) to sites of nerve injury in a rat model of bilateral cavernous nerve injury (BCNI). The effects of sildenafil, tadalafil, and vardenafil on cyclic adenosine monophosphate, cyclic guanosine monophosphate, and cell survival after exposure to hypoxia and H Viability of neuronal cells was measured. Intracavernous pressure changes after cavernous nerve electrostimulation and expression of neurofilament, nitric oxide synthase, and actin in mid-shaft of penis were analyzed 14 days after injury. Sildenafil and rolipram significantly decreased cell death after exposure to H Sildenafil showed the most profound neuroprotective effect compared with tadalafil and vardenafil. Sildenafil- or rolipram-loaded NSP delivery to the site of nerve injury prevented erectile dysfunction and led to increased neurofilament, nitric oxide synthase, smooth muscle content in rat penile tissue after BCNI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6520
Rolipram, solid, ≥98% (HPLC)
C16H21NO3