Cardiovascular drugs and therapy
Evaluation and Treatment of Endoplasmic Reticulum (ER) Stress in Right Ventricular Dysfunction during Monocrotaline-Induced Rat Pulmonary Arterial Hypertension.
PMID 27844183
Endoplasmic reticulum (ER) stress contributes to pulmonary artery hypertension (PAH). However, the exact roles of ER stress in right ventricular (RV) dysfunction, which is strongly associated with PAH, are largely unknown. Here, we aimed to explore how ER stress affects RV function in a rat PAH model and evaluated the effects of an ER stress inhibitor on RV dysfunction. We examined expression changes of an ER marker: chaperone glucose-regulated protein 78 (GRP78), three ER stress sensor proteins: activating transcription factor 6 (ATF6), inositol-requiring enzyme 1 (IRE1), and protein kinase RNA-like endoplasmic reticulum kinase (PERK), and a key ER stress-induced apoptosis indicator: CCAAT/enhancer-binding protein homologous protein (CHOP), with inflammation indicators: interleukin 6 (IL-6), tumor necrosis factor-α (TNF-α), and matrix metalloproteinases (MMPs) in RV at 3, 7, 14 and 28xa0days following a single dose of monocrotaline (MCT) injection, with or without a preventive treatment [4-phenylbutyric acid (PBA)]. RV function was evaluated by histological, molecular and echocardiographic analysis. 1) GRP78 protein expression started to increase (1.5xa0±xa00.06 fold change) at 3d post MCT injection, even before the formation of PAH. 2) ATF6, IRE1, and PERK showed distinctive expression patterns post MCT injection. 3) CHOP expression remained low at day 3 & 7, but significantly increased at day 14 (pxa0