EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
A novel fluorescence immunoassay for the sensitive detection of Escherichia coli O157:H7 in milk based on catalase-mediated fluorescence quenching of CdTe quantum dots.
PMID 27846989
Abstract
Immunoassay is a powerful tool for rapid detection of food borne pathogens in food safety monitoring. However, conventional immunoassay always suffers from low sensitivity when it employs enzyme-catalyzing chromogenic substrates to generate colored molecules as signal outputs. In the present study, we report a novel fluorescence immunoassay for the sensitive detection of E. coli O157:H7 through combination of the ultrahigh bioactivity of catalase to hydrogen peroxide (H
Related Materials