EMAIL THIS PAGE TO A FRIEND
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association
Validation of the Hong Kong Liver Cancer Staging System in Determining Prognosis of the North American Patients Following Intra-arterial Therapy.
PMID 27847278
Abstract
There is debate over the best way to stage hepatocellular carcinoma (HCC). We attempted to validate the prognostic and clinical utility of the recently developed Hong Kong Liver Cancer (HKLC) staging system, a hepatitis B-based model, and compared data with that from the Barcelona Clinic Liver Cancer (BCLC) staging system in a North American population that underwent intra-arterial therapy (IAT). We performed a retrospective analysis of data from 1009 patients with HCC who underwent IAT from 2000 through 2014. Most patients had hepatitis C or unresectable tumors; all patients underwent IAT, with or without resection, transplantation, and/or systemic chemotherapy. We calculated HCC stage for each patient using 5-stage HKLC (HKLC-5) and 9-stage HKLC (HKLC-9) system classifications, and the BCLC system. Survival information was collected up until the end of 2014 at which point living or unconfirmed patients were censored. We compared performance of the BCLC, HKLC-5, and HKLC-9 systems in predicting patient outcomes using Kaplan-Meier estimates, calibration plots, C statistic, Akaike information criterion, and the likelihood ratio test. Median overall survival time, calculated from first IAT until date of death or censorship, for the entire cohort (all stages) was 9.8 months. The BCLC and HKLC staging systems predicted patient survival times with significance (P < .001). HKLC-5 and HKLC-9 each demonstrated good calibration. The HKLC-5 system outperformed the BCLC system in predicting patient survival times (HKLC C = 0.71, Akaike information criterion = 6242; BCLC C = 0.64, Akaike information criterion = 6320), reducing error in predicting survival time (HKLC reduced error by 14%, BCLC reduced error by 12%), and homogeneity (HKLC chi-square = 201, P < .001; BCLC chi-square = 119, P < .001) and monotonicity (HKLC linear trend chi-square = 193, P < .001; BCLC linear trend chi-square = 111, P < .001). Small proportions of patients with HCC of stages IV or V, according to the HKLC system, survived for 6 months and 4 months, respectively. In a retrospective analysis of patients who underwent IAT for unresectable HCC, we found the HKLC-5 staging system to have the best combination of performances in survival separation, calibration, and discrimination; it consistently outperformed the BCLC system in predicting survival times of patients. The HKLC system identified patients with HCC of stages IV and V who are unlikely to benefit from IAT.