EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Evaluation of functionalized mesoporous silica SBA-15 as a carrier system for Ph
PMID 27847952
Abstract
SBA-15|Sn3, a mesoporous silica-based material (derivative of SBA-15) loaded with an organotin compound Ph
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
569615
(3-Chloropropyl)triethoxysilane, 95%
C9H21ClO3Si