EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Sensitivity to Restimulation-Induced Cell Death Is Linked to Glycolytic Metabolism in Human T Cells.
PMID 27852741
Abstract
Restimulation-induced cell death (RICD) regulates immune responses by restraining effector T cell expansion and limiting nonspecific damage to the host. RICD is triggered by re-engagement of the TCR on a cycling effector T cell, resulting in apoptosis. It remains unclear how RICD sensitivity is calibrated in T cells derived from different individuals or subsets. In this study we show that aerobic glycolysis strongly correlates with RICD sensitivity in human CD8
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75351
Oligomycin A, ≥99% (HPLC)
C45H74O11