EMAIL THIS PAGE TO A FRIEND
Human & experimental toxicology
The vasorelaxant mechanisms of methanol on isolated rat aortic rings: Involvement of ion channels and signal transduction pathways.
PMID 27852936
Abstract
It is reported that methanol is generally used as an industrial solvent, antifreeze, windshield washer fluid, cooking fuel and perfume. Methanol ingestion can lead to severe metabolic disturbances, blindness, or even death. So far, few studies about its negative effects on cardiovascular system have been reported. The purpose of this study was to determine the vasoactive effect of methanol and roles of ion channels and signal transduction pathways on isolated rat aorta. The results suggested that the mechanism of methanol-induced vasorelaxation at low concentrations (<500 mM) was mediated by ATP-sensitive K