EMAIL THIS PAGE TO A FRIEND
The Journal of toxicological sciences
Efficacy and safety of a therapeutic apparatus using hydrogen peroxide photolysis to treat dental and periodontal infectious diseases.
PMID 27853108
Abstract
The present study aimed to evaluate the acute locally injurious property of our most current hydroxyl radical generation system by hydrogen peroxide (H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
115746
2,2,6,6-Tetramethyl-4-piperidinol, 98%
C9H19NO