EMAIL THIS PAGE TO A FRIEND
Scientific reports
Intronless WNT10B-short variant underlies new recurrent allele-specific rearrangement in acute myeloid leukaemia.
PMID 27853307
Abstract
Defects in the control of Wnt signaling have emerged as a recurrent mechanism involved in cancer pathogenesis and acute myeloid leukaemia (AML), including the hematopoietic regeneration-associated WNT10B in AC133
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330825
Manganese(II) acetate, 98%
C4H6MnO4
P0040
PP1, ≥98% (HPLC)
C16H19N5