EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Dispersed oil decreases the ability of a model fish (Dicentrarchus labrax) to cope with hydrostatic pressure.
PMID 27854059
Abstract
Data on the biological impact of oil dispersion in deep-sea environment are scarce. Hence, the aim of this study was to evaluate the potential interest of a pressure challenge as a new experimental approach for the assessment of consequences of chemically dispersed oil, followed by a high hydrostatic pressure challenge. This work was conducted on a model fish: juvenile Dicentrarchus labrax. Seabass were exposed for 48 h to dispersant alone (nominal concentration (NC) = 4 mg L