EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Enhanced Neurogenic Biomarker Expression and Reinnervation in Human Acute Skin Wounds Treated by Electrical Stimulation.
PMID 27856290
Abstract
Electrical stimulation (ES) is known to promote cutaneous healing; however, its ability to regulate reinnervation remains unclear. First, we show that ES treatment of human acute cutaneous wounds (n = 40) increased reinnervation. Next, to define neurophysiologic mechanisms through which ES affects repair, microarray analysis of wound biopsy samples was performed on days 3, 7, 10, and 14 after wounding. This identified neural differentiation biomarkers TUBB3 (melanocyte development and neuronal marker) and its upstream molecule FIG4 (phosphatidylinositol (3,5)-bisphosphate 5-phosphatase) as significantly up-regulated after ES treatment. To demonstrate a functional ES-TUBB3 axis in cutaneous healing, we showed increased TUBB3
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5304 Formalin, 10%, Neutral Buffered with 0.03% Eosin