EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Bacterial Peptide Deformylase Inhibition of Tetrazole-Substituted Biaryl Acid Analogs: Synthesis, Biological Evaluations, and Molecular Docking Study.
PMID 27859538
Abstract
The synthesis and screening of tetrazole-substituted biaryl acid analogs 7a-l as bacterial peptide deformylase (PDF) enzyme inhibitors is reported. The compounds 7e (IC