EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
Structural analysis of oleanane-type saponins from the roots of Wisteria frutescens.
PMID 27859693
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149691
Thiazolidine, 95%
C3H7NS