EMAIL THIS PAGE TO A FRIEND
Clinical endocrinology
Maternal 25-hydroxyvitamin D is inversely correlated with foetal serotonin.
PMID 27862146
Abstract
Maternal vitamin D deficiency during pregnancy has been linked to impaired neurocognitive development in childhood. The mechanism by which vitamin D affects childhood neurocognition is unclear but may be via interactions with serotonin, a neurotransmitter involved in foetal brain development. In this study, we aimed to explore associations between maternal and foetal vitamin D concentrations, and foetal serotonin concentrations at term. Serum 25-hydroxyvitamin D (25(OH)D, nmol/l) and serotonin (5-HT, nmol/l) concentrations were measured in maternal and umbilical cord blood from mother-infant pairs (n = 64). Association between maternal 25(OH)D, cord 25(OH)D and cord serotonin was explored using linear regression, before and after adjusting for maternal serotonin levels. We also assessed the effects of siRNA knockdown of the vitamin D receptor (VDR) and administration of 10 nm 1,25-dihydroxyvitamin D We observed an inverse relationship between both maternal and cord 25(OH)D concentrations with cord serotonin concentrations. The treatment of HUVECs with 1,25-dihydroxyvitamin D These observations provide the first evidence of an inverse relationship between maternal 25(OH)D and foetal serotonin concentrations. We propose that maternal vitamin D deficiency increases foetal serotonin concentrations and thereby contributes to longer-term neurocognitive impairment in infants and children.