EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Polar extracts from the berry-like fruits of Hypericum androsaemum L. as a promising ingredient in skin care formulations.
PMID 27864112
Abstract
The top flowering aerial parts of the Hypericum species are traditionally used to prepare ointments to heal cuts and burns. Sometimes even the fruits are used for these purposes. Hypericum androsaemum L., commonly known as tutsan or shrubby St. John's Wort, is a Mediterranean medicinal plant which has been traditionally used to prepare an ointment for treating cuts and wounds. To evaluate the extracts obtained from H. androsaemum red berries as functional ingredients for skin care formulations. The methanolic extract was obtained by Soxhlet extraction while the aqueous extract was prepared by decoction; their composition was determined by HPLC analysis. Their biological activities were measured in terms of proliferation and migration of human fibroblasts, inhibition of collagenase activity, and immunomodulatory effects on human peripheral blood mononuclear cells (PBMCs). In addition, we evaluated their photostability by UV spectroscopy and their protective effects against APPH-induced hemolysis in red blood cells (RBC). The polar extracts contained significant amounts of shikimic (108,143.7-115,901.3mg/kg) and chlorogenic acids (45,781.1-57,002.7mg/kg). The main components of these extracts made an important contribution to a significant increase in human fibroblast migration. Both extracts were also active as collagenase inhibitors, with the aqueous one showing a greater inhibitory capacity (IC H. androsaemum red berries were proven to contain phytochemicals that improve skin regeneration, hence potentially employable in skin care formulations.