EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Biochemical methane potential of oil-extracted microalgae and glycerol in co-digestion with chicken litter.
PMID 27864134
Abstract
The objective of this work was to evaluate the technical feasibility of using both oil-extracted microalgae (M) and glycerol (G) in co-digestion with chicken litter (CL), thereby improving biochemical methane potential (BMP). Different feedstock ratios of M (0-30%), G (0-3%) and CL (67-100%) were investigated to determine the best co-digestion condition under mesophilic conditions. According to the modified Gompertz model, the best BMP (131.1mLCH