EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Induction of Airway Allergic Inflammation by Hypothiocyanite via Epithelial Cells.
PMID 27864363
Abstract
Hypothiocyanite (OSCN
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3262
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
C10H12N5O6P · C4H11NO3
A6733
Auranofin, ≥98% (HPLC)
C20H34AuO9PS