EMAIL THIS PAGE TO A FRIEND
Lasers in medical science
Low-level laser irradiation modulates brain-derived neurotrophic factor mRNA transcription through calcium-dependent activation of the ERK/CREB pathway.
PMID 27864646
Abstract
Low-level laser (LLL) irradiation has been reported to promote neuronal differentiation, but the mechanism remains unclear. Brain-derived neurotrophic factor (BDNF) has been confirmed to be one of the most important neurotrophic factors because it is critical for the differentiation and survival of neurons during development. Thus, this study aimed to investigate the effects of LLL irradiation on Bdnf messenger RNA (mRNA) transcription and the molecular pathway involved in LLL-induced Bdnf mRNA transcription in cultured dorsal root ganglion neurons (DRGNs) using Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1385
KN-93, ≥98% (HPLC)
C26H29ClN2O4S · H3PO4
T9033
Thapsigargin, ≥98% (HPLC), solid film
C34H50O12
T3830
Triciribine hydrate, ≥97% (HPLC)
C13H16N6O4 · xH2O
X2628
Xestospongin C, film
C28H50N2O2