EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Nitric Oxide and Hydrogen Sulfide Interact When Modulating Gastric Physiological Functions in Rodents.
PMID 27864656
Abstract
The objective was to evaluate the effects of nitric oxide (NO) and hydrogen sulfide (H Mice received saline, sodium nitroprusside (SNP), or sodium hydrosulfite (NaHS), and after 1 h, the animals were killed for immunofluorescence analysis of CSE or eNOS expressions, respectively. Other groups received saline, SNP, NaHS, Lawesson's reagent (H SNP and NaHS increased the expression of CSE or eNOS, respectively. SNP or Lawesson's reagent did not alter gastric acid secretion but increased mucus production, and these effects reverted with PAG and L-NAME treatment, respectively. SNP or NaHS increased gastric blood flow and protected the gastric mucosa against ethanol injury, and these effects reverted with PAG and L-NAME treatments, respectively. SNP delayed gastric emptying when compared with saline, and PAG partially reversed this effect. NaHS accelerate gastric emptying, and L-NAME partially reversed this effect. SNP and NaHS alone induced gastric fundus and pylorus relaxation. However, pretreatment with PAG or L-NAME reversed these relaxant effects only in the pylorus but not in the gastric fundus. NO and H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
227439
Lawesson reagent, 97%
C14H14O2P2S4