EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Layered Post-Transition-Metal Dichalcogenides (X-M-M-X) and Their Properties.
PMID 27865023
Abstract
A
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263273
Gallium, 99.999% trace metals basis
Ga