EMAIL THIS PAGE TO A FRIEND
Cellular immunology
Cannabidiol (CBD) induces functional Tregs in response to low-level T cell activation.
PMID 27865421
Abstract
Many effects of the non-psychoactive cannabinoid, cannabidiol (CBD), have been described in immune responses induced by strong immunological stimuli. It has also been shown that CBD enhances IL-2 production in response to low-level T cell stimulation. Since IL-2, in combination with TGF-β1, are critical for Treg induction, we hypothesized that CBD would induce CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248401
Magnesium methyl carbonate solution, 2.0 M in DMF
C3H6MgO4