EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
The adverse impact of obesity on heart rate variability is modified by a NFE2L2 gene variant: The SAPALDIA cohort.
PMID 27866025
Abstract
Overweight has been associated with an increase in inflammatory markers and with an imbalance in the autonomic nervous system, such as a decrease in heart rate variability (HRV). In this study we aimed to investigate the modifying effect of a genetic variation in a major anti-inflammatory marker gene, NFE2L2, on the relationship between overweight and HRV. We analyzed participants of the SAPALDIA cohort aged 50years and older, twice in 2002/2003 (N=1472) and 2010/2011 (N=1235). We included persons with valid genotype data, who underwent ambulatory 24-h electrocardiogram monitoring, and reported on medical history and lifestyle. The association between HRV and BMI, measured as standard deviation of normal-to-normal intervals (SDNN) by BMI and the modifying effect of the cardiovascular health-related NFE2L2 gene variant rs2364723 were tested, applying multivariable mixed linear regression models. We found study participants with overweight (BMI>25) over two follow-up surveys 10years apart to have a negative association between SDNN, calculated as geometric means, with BMI. The examined NFE2L2 variant sustainably modified (p Our results are consistent with the hypothesis that overweight decreases heart rate variability through inflammatory processes.
Related Materials