EMAIL THIS PAGE TO A FRIEND
Toxicology letters
The anti-hyperplasia, anti-oxidative and anti-inflammatory properties of Qing Ye Dan and swertiamarin in testosterone-induced benign prostatic hyperplasia in rats.
PMID 27866977
Abstract
Qing Ye Dan (QYD) is the whole plant of Swertia mileensis and used in Chinese folk medicine for the treatment of prostatitis, benign prostatic hyperplasia (BPH) and so on. This study was to investigate the effects of QYD and its main component swertiamarin on BPH induced by testosterone in rats. The prostatic expressions of vascular endothelial growth factor (VEGF), epidermal growth factor (EGF), basic fibroblast growth factor (βFGF) and proliferating cell nuclear antigen (PCNA) were detected by immunohistochemistry assay. Prostatic levels of oxidative stress and inflammatory-related factors were also analyzed. Additionally, the prostatic expressions of androgen receptor (AR), estrogen receptor (ER)-α, ER-β, hypoxia-inducible factor (HIF)-1α, B-cell CLL/lymphoma (Bcl)-2 and Bcl-2-associated X protein (Bax) were measured by western blot. The epithelial-mesenchymal transition (EMT) associated factors were evaluated by quantitative RT-PCR. It showed that QYD and swertiamarin ameliorated the testosterone-induced prostatic hyperplasia and collagen deposition, attenuated the over-expressions of HIF-1α, VEGF, EGF, βFGF, PCNA, AR and ER-α, reduced the ratio of Bcl-2/Bax, enhanced the expression of ER-β, inhibited the oxidative stress and local inflammation, as well as relieved prostatic EMT. It suggested that QYD and swertiamarin had prostatic protective potential against BPH.