Enzyme and microbial technology
The family 22 carbohydrate-binding module of bifunctional xylanase/β-glucanase Xyn10E from Paenibacillus curdlanolyticus B-6 has an important role in lignocellulose degradation.
PMID 27871388
A newly isolated endo-β-1,4-xylanase (Xyn10E) from Paenibacillus curdlanolyticus B-6 has a modular structure consisting of a family 22 carbohydrate-binding module (CBM), a glycoside hydrolase (GH) family 10 catalytic domain, two fibronectin type III (Fn3) domains, and a family 3 CBM at the C-terminus. Intact Xyn10E (rXyn10E), CBM22-deleted Xyn10E (X-CBM3), CBM3-deleted Xyn10E (X-CBM22), and GH10 catalytic domain only (X-GH10) were expressed in Escherichia coli. rXyn10E showed bifunctional degradation activity toward xylan and β-glucan and also degraded microcrystalline cellulose. Although X-CBM3 and X-GH10 had drastically reduced xylanase and β-glucanase activities, X-CBM22 mostly retained these activities. Similar Km values were obtained for rXyn10E and X-CBM3, but kcat and kcat/Km values for X-CBM3 and X-GH10 were lower than those for rXyn10E, suggesting that CBM22 of Xyn10E may contribute to catalytic efficiency. In binding assays, X-CBM3 was still able to bind to β-glucan, soluble xylan, insoluble xylan, and cellulose through GH10 and CBM3. These results indicate that CBM22 has an important role not only in binding to xylan and β-glucan but also in feeding both polysaccharides into the neighboring GH10 catalytic domain. rXyn10E showed remarkable synergism with rXyn11A, a major xylanase subunit of P. curdlanolyticus B-6, in the degradation of untreated corn stover and sugarcane bagasse; however, the combination of X-CBM3 and rXyn11A was not synergistic. These results indicate that Xyn10E and Xyn11A act synergistically on lignocellulosic biomass, and CBM22 is essential for efficient degradation of lignocellulosic materials.
Related Materials
Product #
Molecular Formula
Add to Cart
4-Nitrophenyl β-D-xylopyranoside, ≥98%