EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Multidrug and toxin extrusion proteins mediate cellular transport of cadmium.
PMID 27871888
Abstract
Cadmium (Cd) is an environmentally prevalent toxicant posing increasing risk to human health worldwide. As compared to the extensive research in Cd tissue accumulation, little was known about the elimination of Cd, particularly its toxic form, Cd ion (Cd