EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The Major Replicative Histone Chaperone CAF-1 Suppresses the Activity of the DNA Mismatch Repair System in the Cytotoxic Response to a DNA-methylating Agent.
PMID 27872185
Abstract
The DNA mismatch repair (MMR) system corrects DNA mismatches in the genome. It is also required for the cytotoxic response of O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9911
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
C20H16N2O4