EMAIL THIS PAGE TO A FRIEND
Biotechnology for biofuels
Whole-genome de novo sequencing, combined with RNA-Seq analysis, reveals unique genome and physiological features of the amylolytic yeast
PMID 27872659
Abstract
Genomic studies on fungal species with hydrolytic activity have gained increased attention due to their great biotechnological potential for biomass-based biofuel production. The amylolytic yeast We performed whole-genome (WG) de novo sequencing and complete assembly of The completely sequenced