EMAIL THIS PAGE TO A FRIEND
International journal of hematology
Mobilization of human immature hematopoietic progenitors through combinatory use of bortezomib and immunomodulatory drugs.
PMID 27873175
Abstract
Combination use of the proteasome inhibitor bortezomib and the immunomodulatory drugs lenalidomide or thalidomide has provided superior outcomes in multiple myeloma over their single use; however, these combinations can produce significant toxicities. Unexpectedly, we found a small but significant increase in the population of immature granulocytes and erythrocytes/megakaryocytes in peripheral blood in 16 of 22 patients (73%) treated with dexamethasone in combination with bortezomib and immunomodulatory drugs (triplet), but not in any of 25 patients treated with either bortezomib or immunomodulatory drugs with dexamethasone (doublet). These immature cells gradually increased to a peak level (mean 2.6% per white blood cells) with triplet therapy, and disappeared immediately after therapy cessation. The numbers of circulating CD34