EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
Inhalable Ipratropium Bromide Particle Engineering with Multicriteria Optimization.
PMID 27873181
Abstract
Spray-dried ipratropium bromide (IPB) microspheres for oral inhalation were engineered using Quality by Design. The interrogation of material properties, process parameters, and critical product quality attributes interplay enabled rational product design. A 2
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
438243
Tetrapropylammonium chloride, 98%
C12H28ClN