EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Neuroprotective effects of vitexin against isoflurane-induced neurotoxicity by targeting the TRPV1 and NR2B signaling pathways.
PMID 27878303
Abstract
Vitexin is a bioactive compound extracted from hawthorn leaves, which reduces blood pressure and has anti‑inflammatory and potential anticancer effects. However, the mechanisms underlying the protective effects of vitexin against isoflurane‑induced neurotoxicity remain elusive. Therefore, the aim of the present study was to investigate these mechanisms further. Sprague Dawley rats received 1.4% isoflurane in a 100% oxygen environment for 2 h. Human PC12 pheochromocytoma neurosecretory cells were exposed to 2% isoflurane for 12 h before they were treated with 1, 10 or 100 µM vitexin for a further 24 h. Vitexin inhibited the isoflurane-induced cell cytotoxicity and weakened isoflurane-induced neuroinflammation and oxidative stress pathways in PC12 cells. In addition, treatment with vitexin suppressed isoflurane‑induced caspase‑3 activation and increased β-secretase 1 levels in PC12 cells. Furthermore, vitexin treatment decreased the levels of isoflurane‑induced cytosolic calcium and reactive oxygen species, and downregulated the expression of transient receptor potential cation channel subfamily V member 1 (TRPV1) and glutamate ionotropic receptor NMDA type subunit 2B (NR2B) protein expression in isoflurane-treated PC12 cells. These results suggest that vitexin mediates its protective effects against isoflurane-induced neurotoxicity by targeting the TRPV1 and NR2B signaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3262
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
C5H9NO4