EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Endocrinology and metabolism
Glucosamine induces ER stress by disrupting lipid-linked oligosaccharide biosynthesis and N-linked protein glycosylation.
PMID 27879249
Abstract
Glucosamine is an essential substrate for N-linked protein glycosylation. However, elevated levels of glucosamine can induce endoplasmic reticulum (ER) stress. Glucosamine-induced ER stress has been implicated in the development of diabetic complications, including atherosclerosis and hepatic steatosis. In this study, we investigate the potential relationship between the effects of glucosamine on lipid-linked oligosaccharide (LLO) biosynthesis, N-linked glycosylation, and ER homeostasis. Mouse embryonic fibroblasts (MEFs) were cultured in the presence of 0-5 mM glucosamine for up to 18 h, and LLO biosynthesis was monitored by fluorescence-assisted carbohydrate electrophoresis. ER stress was determined by quantification of unfolded protein response (UPR) gene expression. We found that exposure of MEFs to ≥1 mM glucosamine significantly impaired the biosynthesis of mature (Glc
Related Materials