EMAIL THIS PAGE TO A FRIEND
Scientific reports
Naturally Occurring Fc-Dependent Antibody From HIV-Seronegative Individuals Promotes HIV-Induced IFN-α Production.
PMID 27881846
Abstract
A majority of adults without HIV infection and with a low risk of HIV-exposure have plasma IgG antibodies that enhance the rate and magnitude of HIV-induced interferon alpha (IFN-α) production. Fc-dependent IgG-HIV complexes induce IFN-α rapidly and in high titers in response to HIV concentrations that are too low to otherwise stimulate an effective IFN-α response. IFN-α promoting antibody (IPA) counters HIV-specific inhibition of IFN-α production, and compensates for the inherent delay in IFN-α production common to HIV infection and other viruses. Naturally occurring IPA has the potential to initiate a potent IFN-α response early in the course of HIV mucosal invasion in time to terminate infection prior to the creation of a pool of persistently infected cells. The current study adds IPA as a mediator of an Fc-dependent antiviral state capable of preventing HIV infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
693839
6-Phosphonohexanoic acid, 97%
C6H13O5P