EMAIL THIS PAGE TO A FRIEND
Hypertension research : official journal of the Japanese Society of Hypertension
Epigenetic regulation of L-type voltage-gated Ca
PMID 27881847
Abstract
Accumulating evidence has shown that epigenetic regulation is involved in hypertension and aging. L-type voltage-gated Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307823
Sodium hydrogen sulfate, technical grade
HNaO4S